missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-Chloronicotinate 97.0+%, TCI America™

Catalog No. E08525G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08525G 5 g
E085225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08525G Supplier TCI America Supplier No. E08525G

Chemical Identifiers

CAS 49608-01-7
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.61
MDL Number MFCD00082739
InChI Key ILDJJTQWIZLGPO-UHFFFAOYSA-N
Synonym ethyl 6-chloronicotinate, ethyl 6-chloro-3-pyridinecarboxylate, ethyl6-chloronicotinate, 6-chloro-nicotinic acid ethyl ester, 6-chloronicotinic acid ethyl ester, ethyl 6-chloro nicotinate, ethyl 2-chloro-5-pyridinecarboxylate, 3-pyridinecarboxylic acid, 6-chloro-, ethyl ester, pubchem1415, pubchem15179
PubChem CID 2799611
IUPAC Name ethyl 6-chloropyridine-3-carboxylate
SMILES CCOC(=O)C1=CC=C(Cl)N=C1

Specifications

Color White-Yellow
Boiling Point 150°C
Quantity 5 g
Formula Weight 185.61
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Ethyl 6-Chloronicotinate