missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-chloronicotinate, 97%

Catalog No. AAA1472314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1472314 25 g
AAA1472306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1472314 Supplier Thermo Scientific Chemicals Supplier No. A1472314

CAS: 49608-01-7 | C8H8ClNO2 | 185.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 49608-01-7
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.61
MDL Number MFCD00082739
InChI Key ILDJJTQWIZLGPO-UHFFFAOYSA-N
PubChem CID 2799611
IUPAC Name ethyl 6-chloropyridine-3-carboxylate
SMILES CCOC(=O)C1=CC=C(Cl)N=C1

Specifications

Melting Point 26°C to 30°C
Boiling Point 86°C to 87°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.522
Quantity 25 g
Beilstein 4742175
Synonym ethyl 6-chloronicotinate, ethyl 6-chloro-3-pyridinecarboxylate, ethyl6-chloronicotinate, 6-chloro-nicotinic acid ethyl ester, 6-chloronicotinic acid ethyl ester, ethyl 6-chloro nicotinate, ethyl 2-chloro-5-pyridinecarboxylate, 3-pyridinecarboxylic acid, 6-chloro-, ethyl ester, pubchem1415, pubchem15179
Formula Weight 185.61
Percent Purity 97%
Chemical Name or Material Ethyl 6-chloronicotinate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only