accessibility menu, dialog, popup

UserName

HN

CAS RN 496-15-1

IUPAC Name: 2,3-dihydro-1H-indole
Synonyms: indoline dihydroindole
Molecular Weight (g/mol): 119.17
Molecular Formula: C8H9N
InChi Key: LPAGFVYQRIESJQ-UHFFFAOYSA-N
SMILES: C1CC2=CC=CC=C2N1
Boiling Point 
  • (6)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (6)
  • (2)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)