accessibility menu, dialog, popup

UserName

H3CO

CAS RN 495-40-9

IUPAC Name: 1-phenylbutan-1-one
Synonyms: butyrophenone
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: FFSAXUULYPJSKH-UHFFFAOYSA-N
SMILES: CCCC(=O)C1=CC=CC=C1
Percent Purity 
  • (4)
  • (5)
Boiling Point 
  • (3)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (5)
  • (4)
  • (5)
  • (3)
  • (3)
  • (2)
  • (2)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (1)