missing translation for 'onlineSavingsMsg'
Learn More

Butyrophenone, Analytical Standard, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014317
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014317 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014317 Supplier Merck Emd Millipore Supplier No. 167041ML

Specifications

CAS 495-40-9
Density 1.021 g/mL (at 25°C)
Linear Formula C6H5COCH2CH2CH3
MDL Number MFCD00009397
Melting Point 11°C to 13°C
Boiling Point 228°C to 230°C
Synonym 1-Benzoylpropane; 1-Phenyl-1-butanone; NSC 8463; Phenyl propyl ketone
Quantity 1 mL
Formula Weight 148.20
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less