missing translation for 'onlineSavingsMsg'
Learn More

Butyrophenone, 99%

Catalog No. AAA1134530
Change view
Click to view available options
Quantity:
25 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1134530 250 g
AAA1134514 25 g
AAA1134518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1134530 Supplier Thermo Scientific Chemicals Supplier No. A1134530
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 495-40-9 | C10H12O | 148.205 g/mol

Butyrophenone is used as intermediates of Liquid Crystals

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Butyrophenone is used as intermediates of Liquid Crystals

Solubility
Insoluble in water.

Notes
Protect from light. Store under an inert atmosphere.

Chemical Identifiers

CAS 495-40-9
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00009397
InChI Key FFSAXUULYPJSKH-UHFFFAOYSA-N
Synonym butyrophenone, n-butyrophenone, 1-butanone, 1-phenyl, phenyl propyl ketone, 1-phenyl-1-butanone, propyl phenyl ketone, unii-186i11wb5b, 1-phenyl-butan-1-one, propylphenylketone
PubChem CID 10315
IUPAC Name 1-phenylbutan-1-one
SMILES CCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 11°C to 13°C
Density 1.021
Boiling Point 222°C
Flash Point 88°C (190°F)
Odor Characteristic
Refractive Index 1.5195
Quantity 250 g
Beilstein 508305
Solubility Information Insoluble in water.
Formula Weight 148.21
Percent Purity 99%
Chemical Name or Material Butyrophenone
Show More Show Less
Hazard Category H227
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P370+P378q-P374-P501c
EINECSNumber 207-799-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only