accessibility menu, dialog, popup

UserName

CAS RN 4949-69-3

CH3NH2I

CAS RN 4949-69-3

IUPAC Name: 4-iodo-3-methylaniline
Synonyms: 4-iodo-3-methylaniline
Molecular Weight (g/mol): 233.05
Molecular Formula: C7H8IN
InChi Key: UISBOJCPTKUBIC-UHFFFAOYSA-N
SMILES: CC1=CC(N)=CC=C1I
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)