accessibility menu, dialog, popup

UserName

CH3H3CHOOO

CAS RN 4919-37-3

IUPAC Name: 4-hydroxy-3,5-dimethylbenzoate
Synonyms: 4-hydroxy-3,5-dimethylbenzoate
Molecular Weight (g/mol): 165.17
Molecular Formula: C9H9O3
InChi Key: OMNHTTWQSSUZHO-UHFFFAOYSA-M
SMILES: CC1=CC(=CC(C)=C1O)C([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)