accessibility menu, dialog, popup

UserName

CAS RN 491-80-5

IUPAC Name: 5,7-dihydroxy-3-(4-methoxyphenyl)-4H-chromen-4-one
Synonyms: biochanin biochanin A, 14C-labeled genistein 4-methylether genistein 4-methyl ether biochanine a pratensol biochanin a biochanin A 14C-labeled biochanin A olmelin
Molecular Weight (g/mol): 284.27
Molecular Formula: C16H12O5
InChi Key: WUADCCWRTIWANL-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=COC2=CC(O)=CC(O)=C2C1=O
Percent Purity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)