accessibility menu, dialog, popup

UserName

HOOOOHHO

CAS RN 491-67-8

IUPAC Name: 5,6,7-trihydroxy-2-phenyl-4H-chromen-4-one
Synonyms: baicalein noroxylin biacalein
Molecular Weight (g/mol): 270.24
Molecular Formula: C15H10O5
InChi Key: FXNFHKRTJBSTCS-UHFFFAOYSA-N
SMILES: OC1=C(O)C(O)=C2C(=O)C=C(OC2=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)