missing translation for 'onlineSavingsMsg'
Learn More

Baicalein, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014308
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014308 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014308 Supplier Merck Emd Millipore Supplier No. 9208150MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity phytochemical standard for analytical applications.

Specifications

Chemical Name or Material Baicalein
CAS 491-67-8
Linear Formula C15H10O5
MDL Number MFCD00017459
Melting Point 256°C to 271°C
Synonym 5,6,7-Trihydroxyflavone
Type Analytical Standard
Industry Type Laboratory Use
Molecular Weight (g/mol) 270.24
Quantity 50 mg
Formula Weight 270.24
Percent Purity ≥95% (HPLC)
Grade Analytical Standard
Show More Show Less