missing translation for 'onlineSavingsMsg'
Learn More

Baicalein 98.0+%, TCI America™

Catalog No. T27215G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27215G 5 g
T27211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27215G Supplier TCI America Supplier No. T27215G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 491-67-8
Molecular Formula C15H10O5
Molecular Weight (g/mol) 270.24
MDL Number MFCD00017459
InChI Key FXNFHKRTJBSTCS-UHFFFAOYSA-N
Synonym baicalein, 5,6,7-trihydroxyflavone, noroxylin, 5,6,7-trihydroxy-2-phenyl-4h-chromen-4-one, biacalein, baicelein, chembl8260, 4h-1-benzopyran-4-one, 5,6,7-trihydroxy-2-phenyl, 5,7-trihydroxyflavone, 5,6,7-trihydroxy-2-phenyl-chromen-4-one
PubChem CID 5281605
ChEBI CHEBI:2979
IUPAC Name 5,6,7-trihydroxy-2-phenyl-4H-chromen-4-one
SMILES OC1=C(O)C(O)=C2C(=O)C=C(OC2=C1)C1=CC=CC=C1

Specifications

Melting Point 265°C
Color Yellow
Quantity 5 g
Formula Weight 270.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Baicalein
RTECSNumber DJ3100898
TSCA No
Recommended Storage Refrigerator