CAS RN 4897-25-0
5-Chloro-1-Methyl-4-Nitroimidazole, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™
BP Reference standard, intended for use in laboratory tests.
5-Chloro-1-methyl-4-nitroimidazole, 95%
CAS: 4897-25-0 Molecular Formula: C4H4ClN3O2 Molecular Weight (g/mol): 161.545 MDL Number: MFCD00233664 InChI Key: OSJUNMSWBBOTQU-UHFFFAOYSA-N Synonym: 5-chloro-1-methyl-4-nitro-1h-imidazole,1h-imidazole, 5-chloro-1-methyl-4-nitro,1-methyl-5-chloro-4-nitroimidazole,pcmni,imidazole, 5-chloro-1-methyl-4-nitro,unii-p917jw98bd,1-methyl-4-nitro-5-chloroimidazole,1-methyl-4-nitro-5-chloro imidazole,cmni,acmc-1api8 PubChem CID: 21010 IUPAC Name: 5-chloro-1-methyl-4-nitroimidazole SMILES: CN1C=NC(=C1Cl)[N+](=O)[O-]
5-Chloro-1-methyl-4-nitroimidazole 98.0+%, TCI America™
CAS: 4897-25-0 Molecular Formula: C4H4ClN3O2 Molecular Weight (g/mol): 161.545 MDL Number: MFCD00233664 InChI Key: OSJUNMSWBBOTQU-UHFFFAOYSA-N Synonym: 5-chloro-1-methyl-4-nitro-1h-imidazole,1h-imidazole, 5-chloro-1-methyl-4-nitro,1-methyl-5-chloro-4-nitroimidazole,pcmni,imidazole, 5-chloro-1-methyl-4-nitro,unii-p917jw98bd,1-methyl-4-nitro-5-chloroimidazole,1-methyl-4-nitro-5-chloro imidazole,cmni,acmc-1api8 PubChem CID: 21010 IUPAC Name: 5-chloro-1-methyl-4-nitroimidazole SMILES: CN1C=NC(=C1Cl)[N+](=O)[O-]