missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-1-methyl-4-nitroimidazole 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 147°C |
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 161.55 |
| Percent Purity | ≥98.0% (N) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 5-Chloro-1-methyl-4-nitroimidazole |
Safety and Handling
| RTECSNumber | NI4397100 |
| TSCA | No |