missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1-Methyl-4-Nitroimidazole, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022970
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022970 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022970 Supplier Merck Emd Millipore Supplier No. BP965

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BP Reference standard, intended for use in laboratory tests.

Specifications

CAS 4897-25-0
Melting Point 148°C to 150°C (literature)
Molecular Formula C4H4ClN3O2
Linear Formula C4H4ClN3O2
Quantity 100 mg
MDL Number MFCD00233664
Molecular Weight (g/mol) 161.55
Formula Weight 161.55
Grade Not Available
Physical Form Solid