missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1-methyl-4-nitroimidazole, 95%

Catalog No. AAH6182406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6182406 5 g
AAH6182403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6182406 Supplier Thermo Scientific Chemicals Supplier No. H6182406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4897-25-0 | C4H4ClN3O2 | 161.545 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4897-25-0
Molecular Formula C4H4ClN3O2
Molecular Weight (g/mol) 161.545
MDL Number MFCD00233664
InChI Key OSJUNMSWBBOTQU-UHFFFAOYSA-N
Synonym 5-chloro-1-methyl-4-nitro-1h-imidazole, 1h-imidazole, 5-chloro-1-methyl-4-nitro, 1-methyl-5-chloro-4-nitroimidazole, pcmni, imidazole, 5-chloro-1-methyl-4-nitro, unii-p917jw98bd, 1-methyl-4-nitro-5-chloroimidazole, 1-methyl-4-nitro-5-chloro imidazole, cmni, acmc-1api8
PubChem CID 21010
IUPAC Name 5-chloro-1-methyl-4-nitroimidazole
SMILES CN1C=NC(=C1Cl)[N+](=O)[O-]

Specifications

Color White
Quantity 5 g
Formula Weight 161.55
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 5-Chloro-1-methyl-4-nitroimidazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 225-521-2
RTECSNumber NI4397100
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only