accessibility menu, dialog, popup

UserName

CAS RN 4870-65-9

IUPAC Name: 2-bromo-2-phenylacetic acid
Synonyms: bromo(phenyl)acetic acid
Molecular Weight (g/mol): 215.05
Molecular Formula: C8H7BrO2
InChi Key: WAKFRZBXTKUFIW-UHFFFAOYNA-N
SMILES: OC(=O)C(Br)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)