accessibility menu, dialog, popup

UserName

CAS RN 4858-85-9

IUPAC Name: 2,3-dichloropyrazine
Synonyms: 2,3-dichloropyrazine
Molecular Weight (g/mol): 148.97
Molecular Formula: C4H2Cl2N2
InChi Key: MLCNOCRGSBCAGH-UHFFFAOYSA-N
SMILES: ClC1=NC=CN=C1Cl
Boiling Point 
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)