accessibility menu, dialog, popup

UserName

H3COOSH2N

CAS RN 4815-34-3

IUPAC Name: ethyl 2-amino-5-phenylthiophene-3-carboxylate
Synonyms: ethyl 2-amino-5-phenylthiophene-3-carboxylate
Molecular Weight (g/mol): 247.31
Molecular Formula: C13H13NO2S
InChi Key: WIVNPGXPJBBZQH-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)SC(=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)