accessibility menu, dialog, popup

UserName

CAS RN 479-27-6

IUPAC Name: naphthalene-1,8-diamine
Synonyms: 1,8-diaminonaphthalene
Molecular Weight (g/mol): 158.2
Molecular Formula: C10H10N2
InChi Key: YFOOEYJGMMJJLS-UHFFFAOYSA-N
SMILES: NC1=CC=CC2=CC=CC(N)=C12
Boiling Point 
  • (3)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (6)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)