missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Rhein, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002078
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002078 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002078 Supplier Merck Emd Millipore Supplier No. PHL8938520MG

missing translation for 'validMainframeMsg'

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 478-43-3
Product Line phyproof
Linear Formula C15H8O6
MDL Number MFCD00009618
Melting Point >=300°C
Synonym 4,5-Dihydroxyanthraquinone-2-carboxylic acid; 9,10-Dihydro-4,5-dihydroxy-9,10-dioxo-2-anthracenecarboxylic acid; Cassic acid; Chrysazin 3-carboxylic acid; Monorhein; Rheic acid; Rhubarb yellow
Type Reference Substance
Quantity 20 mg
Formula Weight 284.22
Percent Purity ≥95% (HPLC)
Grade Primary Reference Standard
Show More Show Less

phyproof is a registered trademark of PhytoLab GmbH & Co. KG