accessibility menu, dialog, popup

UserName

Br

CAS RN 474688-76-1

IUPAC Name: 2-bromo-9,10-bis(naphthalen-2-yl)anthracene
Synonyms: 2-bromo-9,10-bis(naphthalen-2-yl)anthracene
Molecular Weight (g/mol): 509.45
Molecular Formula: C34H21Br
InChi Key: NNVPXSAMRFIMLP-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C(=C1)C(C1=CC=C3C=CC=CC3=C1)=C1C=CC=CC1=C2C1=CC2=CC=CC=C2C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results