accessibility menu, dialog, popup

UserName

CAS RN 4724-47-4

IUPAC Name: octadecylphosphonic acid
Synonyms: octadecylphosphonic acid
Molecular Weight (g/mol): 334.48
Molecular Formula: C18H39O3P
InChi Key: FTMKAMVLFVRZQX-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCP(O)(O)=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)