accessibility menu, dialog, popup

UserName

NaNaH3CH3CS(R)(R)HNO(R)OOSON(S)OO

CAS RN 4697-14-7

IUPAC Name: disodium (2S,5R,6R)-6-[(2R)-2-carboxylato-2-(thiophen-3-yl)acetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Synonyms: ticar ticarcillin disodium ticarcillin sodium ticarpen aerugipen
Molecular Weight (g/mol): 428.38
Molecular Formula: C15H14N2Na2O6S2
InChi Key: ZBBCUBMBMZNEME-QBGWIPKPSA-L
SMILES: [Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](C([O-])=O)C3=CSC=C3)C(=O)N2[C@H]1C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results