accessibility menu, dialog, popup

UserName

SOOO

CAS RN 468751-38-4

IUPAC Name: 2-[(benzenesulfonyl)methyl]benzaldehyde
Synonyms: 2-[(benzenesulfonyl)methyl]benzaldehyde
Molecular Weight (g/mol): 260.31
Molecular Formula: C14H12O3S
InChi Key: KDTVRUZABJBZKA-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC=C1CS(=O)(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results