accessibility menu, dialog, popup

UserName

CAS RN 4669-06-1

NH

CAS RN 4669-06-1

IUPAC Name: N-(naphthalen-1-yl)naphthalen-2-amine
Synonyms: N-(naphthalen-1-yl)naphthalen-2-amine
Molecular Weight (g/mol): 269.35
Molecular Formula: C20H15N
InChi Key: UNJZLNFHHINVOB-UHFFFAOYSA-N
SMILES: N(C1=CC=C2C=CC=CC2=C1)C1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results