accessibility menu, dialog, popup

UserName

CAS RN 464192-28-7

IUPAC Name: 2-bromo-1,3-thiazole-5-carbaldehyde
Synonyms: 2-bromo-1,3-thiazole-5-carbaldehyde
Molecular Weight (g/mol): 192.03
Molecular Formula: C4H2BrNOS
InChi Key: DJUWIZUEHXRECB-UHFFFAOYSA-N
SMILES: BrC1=NC=C(S1)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results