accessibility menu, dialog, popup

UserName

CAS RN 462-06-6

IUPAC Name: fluorobenzene
Synonyms: fluorobenzene
Molecular Weight (g/mol): 96.1
Molecular Formula: C6H5F
InChi Key: PYLWMHQQBFSUBP-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1
Boiling Point 
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (6)