missing translation for 'onlineSavingsMsg'
Learn More

Fluorobenzene 99.0+%, TCI America™

Catalog No. F0034500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0034500G 500 g
F003425G 25 g
F0034100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F0034500G Supplier TCI America Supplier No. F0034500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 462-06-6
Molecular Formula C6H5F
Molecular Weight (g/mol) 96.10
MDL Number MFCD00000280
InChI Key PYLWMHQQBFSUBP-UHFFFAOYSA-N
Synonym benzene, fluoro, monofluorobenzene, phenyl fluoride, fluorbenzol, fluoro-benzene, 4-fluorobenzene, unii-g3tsz68k12, fluoranylbenzene, fluorobenzenes, 1,2,3,4,5-pentadeuterio-6-fluorobenzene
PubChem CID 10008
ChEBI CHEBI:5115
IUPAC Name fluorobenzene
SMILES FC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 85°C
Quantity 500 g
UN Number 2387
Formula Weight 96.10
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Fluorobenzene
EINECSNumber (9)-1232
RTECSNumber DA0800000
TSCA Yes