accessibility menu, dialog, popup

UserName

H3COHO

CAS RN 4521-22-6

IUPAC Name: 4-(4-methylphenyl)butanoic acid
Synonyms: 4-(4-methylphenyl)butanoic acid
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: IXWOVMRDYFFXGI-UHFFFAOYSA-N
SMILES: CC1=CC=C(CCCC(O)=O)C=C1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results