accessibility menu, dialog, popup

UserName

CAS RN 452-86-8

IUPAC Name: 4-methylbenzene-1,2-diol
Synonyms: 4-methyl-1,2-benzenediol 4-metylcatechol 4-methylcatehol homopyrocatechol homocatechol
Molecular Weight (g/mol): 124.14
Molecular Formula: C7H8O2
InChi Key: ZBCATMYQYDCTIZ-UHFFFAOYSA-N
SMILES: CC1=CC=C(O)C(O)=C1
Boiling Point 
  • (2)
  • (6)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (6)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)