accessibility menu, dialog, popup

UserName

CH3FH2N

CAS RN 452-71-1

IUPAC Name: 4-fluoro-2-methylaniline
Synonyms: 4-fluoro-2-methylaniline
Molecular Weight (g/mol): 125.15
Molecular Formula: C7H8FN
InChi Key: KMHLGVTVACLEJE-UHFFFAOYSA-N
SMILES: CC1=CC(F)=CC=C1N
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)