accessibility menu, dialog, popup

UserName

CAS RN 4511-42-6

IUPAC Name: 3,6-dimethyl-1,4-dioxane-2,5-dione
Synonyms: lactide dilactide
Molecular Weight (g/mol): 144.13
Molecular Formula: C6H8O4
InChi Key: JJTUDXZGHPGLLC-UHFFFAOYNA-N
SMILES: CC1OC(=O)C(C)OC1=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)