accessibility menu, dialog, popup

UserName

H3CO

CAS RN 4466-24-4

IUPAC Name: 2-butylfuran
Synonyms: 2-butylfuran
Molecular Weight (g/mol): 124.18
Molecular Formula: C8H12O
InChi Key: NWZIYQNUCXUJJJ-UHFFFAOYSA-N
SMILES: CCCCC1=CC=CO1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)