missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-n-Butylfuran, 98%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| CAS | 4466-24-4 |
| Density | 0.886 |
| Boiling Point | 153°C to 154°C |
| Flash Point | 32°C (89°F) |
| Odor | Pleasant |
| Molecular Formula | C8H12O |
| Quantity | 50g |
| MDL Number | MFCD00047071 |
| UN Number | UN1993 |
| Beilstein | 107045 |
| Show More |
Safety and Handling
| Hazard Category | H226 |
| Hazard Statement | GHS H Statement H226 Flammable liquid and vapour. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 224-732-7 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only