missing translation for 'onlineSavingsMsg'
Learn More

2-Butylfuran 98.0+%, TCI America™

Catalog No. B24125G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B24125G 5 g
B241225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B24125G Supplier TCI America Supplier No. B24125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4466-24-4
Molecular Formula C8H12O
Molecular Weight (g/mol) 124.183
MDL Number MFCD00047071
InChI Key NWZIYQNUCXUJJJ-UHFFFAOYSA-N
Synonym furan, 2-butyl, 2-n-butyl furan, 2-n-butylfuran, unii-81jv9zyk0d, 81jv9zyk0d, 2-butyl furan, 2-butyl-furan, 2-but-1-yl fura, 2-but-1-yl furan, 1-fur-2-yl butane
PubChem CID 20534
IUPAC Name 2-butylfuran
SMILES CCCCC1=CC=CO1

Specifications

Color Yellow
Boiling Point 59°C
Quantity 5 g
UN Number 1993
Formula Weight 124.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Butylfuran