CAS RN 446044-45-7
IUPAC Name:
2-{1,3-dioxo-1H,2H,3H-naphtho[2,3-f]isoindol-2-yl}cyclohexane-1-carboxylic acid
Synonyms:
2-{1,3-dioxonaphtho[2,3-f]isoindol-2-yl}cyclohexane-1-carboxylic acid
Molecular Weight (g/mol):
373.41
Molecular Formula:
C23H19NO4
InChi Key:
IIJKRPVAVXANME-UHFFFAOYNA-N
SMILES:
OC(=O)C1CCCCC1N1C(=O)C2=C(C=C3C=C4C=CC=CC4=CC3=C2)C1=O
1
–
1
of
1
results
1
(1S,2S)-2-(Anthracene-2,3-dicarboximido)cyclohexanecarboxylic Acid 97.0+%, TCI America™
CAS: 446044-45-7 Molecular Formula: C23H19NO4 Molecular Weight (g/mol): 373.408 InChI Key: IIJKRPVAVXANME-PXNSSMCTSA-N Synonym: N-[(1S,2S)-2-Carboxycyclohexyl]anthracene-2,3-dicarboximide PubChem CID: 44629755 IUPAC Name: (1S,2S)-2-(1,3-dioxonaphtho[2,3-f]isoindol-2-yl)cyclohexane-1-carboxylic acid SMILES: C1CCC(C(C1)C(=O)O)N2C(=O)C3=CC4=CC5=CC=CC=C5C=C4C=C3C2=O