accessibility menu, dialog, popup

UserName

CAS RN 446-11-7

IUPAC Name: 1-fluoro-4-methyl-2-nitrobenzene
Synonyms: 1-fluoro-4-methyl-2-nitrobenzene
Molecular Weight (g/mol): 155.13
Molecular Formula: C7H6FNO2
InChi Key: OORBDHOQLZRIQR-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C(=C1)[N+]([O-])=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)