missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-nitrotoluene 98.0+%, TCI America™

Catalog No. F05625G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05625G 5 g
F056225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05625G Supplier TCI America Supplier No. F05625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 446-11-7
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD00007060
InChI Key OORBDHOQLZRIQR-UHFFFAOYSA-N
Synonym 4-fluoro-3-nitrotoluene, 3-nitro-4-fluorotoluene, 2-fluoro-5-methylnitrobenzene, 4-methyl-2-nitro-1-fluorobenzene, 1-fluoro-4-methyl-2-nitro-benzene, benzene, 1-fluoro-4-methyl-2-nitro, pubchem1597, acmc-1aqwi, 4-fluro-3-nitrotoluene, 4-fluoro-3-nitro toluene
PubChem CID 136287
IUPAC Name 1-fluoro-4-methyl-2-nitrobenzene
SMILES CC1=CC(=C(C=C1)F)[N+](=O)[O-]

Specifications

Color Yellow
Boiling Point 241°C
Quantity 5 g
Formula Weight 155.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Fluoro-3-nitrotoluene