missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-nitrotoluene, 98%

Catalog No. AAH6331722
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6331722 100 g
AAH6331714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6331722 Supplier Thermo Scientific Chemicals Supplier No. H6331722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 446-11-7 | C7H6FNO2 | 155.128 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 446-11-7
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD00007060
InChI Key OORBDHOQLZRIQR-UHFFFAOYSA-N
Synonym 4-fluoro-3-nitrotoluene, 3-nitro-4-fluorotoluene, 2-fluoro-5-methylnitrobenzene, 4-methyl-2-nitro-1-fluorobenzene, 1-fluoro-4-methyl-2-nitro-benzene, benzene, 1-fluoro-4-methyl-2-nitro, pubchem1597, acmc-1aqwi, 4-fluro-3-nitrotoluene, 4-fluoro-3-nitro toluene
PubChem CID 136287
IUPAC Name 1-fluoro-4-methyl-2-nitrobenzene
SMILES CC1=CC(=C(C=C1)F)[N+](=O)[O-]

Specifications

Color Red to Brown
Boiling Point 241°C (decomposition)
Quantity 100 g
UN Number UN2810
Beilstein 2253853
Formula Weight 155.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Fluoro-3-nitrotoluene
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only