accessibility menu, dialog, popup

UserName

CAS RN 4457-32-3

IUPAC Name: (4-nitrophenyl)methyl carbonochloridate
Synonyms: (4-nitrophenyl)methyl carbonochloridate
Molecular Weight (g/mol): 215.59
Molecular Formula: C8H6ClNO4
InChi Key: MHSGOABISYIYKP-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(COC(Cl)=O)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)