accessibility menu, dialog, popup

UserName

CH3H3COBOH3CH3CClN

CAS RN 444120-94-9

IUPAC Name: 2-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 2-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 239.51
Molecular Formula: C11H15BClNO2
InChi Key: IEFBTHROULVUAH-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(Cl)N=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results