accessibility menu, dialog, popup

UserName

OOO

CAS RN 4430-15-3

IUPAC Name: heptacyclo[18.7.0.0²,¹⁰.0³,⁸.0¹¹,¹⁹.0¹²,¹⁷.0²¹,²⁶]heptacosa-1(20),2(10),3,5,7,11(19),12,14,16,21(26),22,24-dodecaene-9,18,27-trione
Synonyms: heptacyclo[18.7.0.0²,¹⁰.0³,⁸.0¹¹,¹⁹.0¹²,¹⁷.0²¹,²⁶]heptacosa-1(20),2(10),3,5,7,11(19),12,14,16,21(26),22,24-dodecaene-9,18,27-trione
Molecular Weight (g/mol): 384.39
Molecular Formula: C27H12O3
InChi Key: HSMDIUMGCHZHNI-UHFFFAOYSA-N
SMILES: O=C1C2=CC=CC=C2C2=C1C1=C(C(=O)C3=CC=CC=C13)C1=C2C(=O)C2=C1C=CC=C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results