accessibility menu, dialog, popup

UserName

CH3FH3C

CAS RN 443-82-3

IUPAC Name: 1-fluoro-2,3-dimethylbenzene
Synonyms: 1-fluoro-2,3-dimethylbenzene
Molecular Weight (g/mol): 124.16
Molecular Formula: C8H9F
InChi Key: AWLDSXJCQWTJPC-UHFFFAOYSA-N
SMILES: CC1=CC=CC(F)=C1C
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)