missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-o-xylene 98.0+%, TCI America™

Catalog No. F03325ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03325ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F03325ML Supplier TCI America Supplier No. F03325ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity, flammable 3-Fluoro-o-xylene for laboratory research applications.

Chemical Identifiers

CAS 443-82-3
Molecular Formula C8H9F
Molecular Weight (g/mol) 124.16
MDL Number MFCD00000323
InChI Key AWLDSXJCQWTJPC-UHFFFAOYSA-N
Synonym 3-fluoro-o-xylene, 2,3-dimethylfluorobenzene, 3-fluoro-1,2-xylene, o-xylene, 3-fluoro, 3-fluoro-1,2-dimethylbenzene, benzene, 1-fluoro-2,3-dimethyl, 1,2-dimethyl-3-fluorobenzene, 1-fluoro-2,3-dimethyl-benzene, fluorxylol, pubchem4408
PubChem CID 96489
IUPAC Name 1-fluoro-2,3-dimethylbenzene
SMILES CC1=CC=CC(F)=C1C

Specifications

Color Yellow
Quantity 5 mL
Physical Form Clear Liquid at 20°C
UN Number 1993
Boiling Point 148°C
Formula Weight 124.16
Percent Purity ≥98.0% (GC)
Grade Not Available
Chemical Name or Material 3-Fluoro-o-xylene
RTECSNumber ZE4560000
TSCA No