missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-o-xylene, 99%

Catalog No. AAA1063722
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1063722 100 g
AAA1063706 5 g
AAA1063714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1063722 Supplier Thermo Scientific Chemicals Supplier No. A1063722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 443-82-3 | C8H9F | 124.16 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Stable under normal conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 443-82-3
Molecular Formula C8H9F
Molecular Weight (g/mol) 124.16
MDL Number MFCD00000323
InChI Key AWLDSXJCQWTJPC-UHFFFAOYSA-N
Synonym 3-fluoro-o-xylene, 2,3-dimethylfluorobenzene, 3-fluoro-1,2-xylene, o-xylene, 3-fluoro, 3-fluoro-1,2-dimethylbenzene, benzene, 1-fluoro-2,3-dimethyl, 1,2-dimethyl-3-fluorobenzene, 1-fluoro-2,3-dimethyl-benzene, fluorxylol, pubchem4408
PubChem CID 96489
IUPAC Name 1-fluoro-2,3-dimethylbenzene
SMILES CC1=CC=CC(F)=C1C

Specifications

Density 0.992
Quantity 100 g
Odor Aromatic
Refractive Index 1.486
UN Number UN1993
Beilstein 2040955
Boiling Point 147°C to 148°C
Solubility Information Insoluble in water.
Formula Weight 124.16
Percent Purity 99%
Chemical Name or Material 3-Fluoro-o-xylene
Show More Show Less
Hazard Category H226-H335
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 207-140-3
RTECSNumber ZE4560000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only