accessibility menu, dialog, popup

UserName

CAS RN 4373-60-8

IUPAC Name: 2-(3-bromophenyl)pyridine
Synonyms: 2-(3-bromophenyl)pyridine
Molecular Weight (g/mol): 234.1
Molecular Formula: C11H8BrN
InChi Key: WLPFTJXVEBANAM-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=C1)C1=CC=CC=N1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)