accessibility menu, dialog, popup

UserName

CAS RN 4369-08-8

CH3OCH3HO

CAS RN 4369-08-8

IUPAC Name: 1-(4-methylphenoxy)propan-2-ol
Synonyms: 1-(4-methylphenoxy)propan-2-ol
Molecular Weight (g/mol): 166.22
Molecular Formula: C10H14O2
InChi Key: VTOVIJQMFYQNSX-UHFFFAOYNA-N
SMILES: CC(O)COC1=CC=C(C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results