accessibility menu, dialog, popup

UserName

CAS RN 4333-56-6

IUPAC Name: bromocyclopropane
Synonyms: bromocyclopropane
Molecular Weight (g/mol): 120.98
Molecular Formula: C3H5Br
InChi Key: LKXYJYDRLBPHRS-UHFFFAOYSA-N
SMILES: BrC1CC1
Molecular Weight (g/mol) 
  • (6)
  • (2)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (6)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (4)