accessibility menu, dialog, popup

UserName

CAS RN 4333-21-5

ONSN

CAS RN 4333-21-5

IUPAC Name: 2-phenyl-3-sulfanylidene-hexahydro-1H-pyrrolo[1,2-c]imidazol-1-one
Synonyms: 2-phenyl-3-sulfanylidene-tetrahydropyrrolo[1,2-c]imidazol-1-one
Molecular Weight (g/mol): 232.3
Molecular Formula: C12H12N2OS
InChi Key: YNJJBMAKBUZCKN-UHFFFAOYNA-N
SMILES: O=C1C2CCCN2C(=S)N1C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results